Learn More
IgG2a Kpa ItCl Mouse, R718, Clone: G155-178, Isotype Control, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566949
Catalog No. BDB566949
Add to cart
Description
Description
The G155-178 clone has an unknown specificity. Trinitrophenol (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to immunoglobulin Fc receptors. The immunoglobulin secreted by the G155-178 hybridoma was selected as a mouse IgG2a, κ isotype control following screening for low background binding on a variety of mouse and human tissues.
Specifications
Specifications
IgG2a Kpa ItCl
G155-178
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Isotype Control