Learn More
IgG2a Kpa ItCl Rat, R718, Clone: R35-95, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566941
Catalog No. BDB566941
Add to cart
Description
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.
Specifications
Specifications
IgG2a Kpa ItCl
R35-95
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Rat
RUO
Isotype Control