Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a, κ, Mouse, Alexa Fluor 488, Clone: G155-178, Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Supplier: BD Biosciences 565358
Description
IgG2a, κ Monoclonal antibody specifically detects IgG2a, κ in samples. It is validated for Isotype control, Flow cytometry.Specifications
IgG2a, κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Cell Biology | |
Purified |
Flow Cytometry | |
G155-178 | |
Alexa Fluor 488 | |
Isotype Control; Isotype control (unknown) | |
TNP-keyhole limpet hemocyanin | |
RUO | |
Primary | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction