Learn More
IgG2a, κ, Mouse, Alexa Fluor 647, Clone: G155-178, Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Manufacturer:  BD Biosciences 565357
Catalog No. BDB565357
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG2a, κ Monoclonal antibody specifically detects IgG2a, κ in samples. It is validated for Isotype control, Flow cytometry.
Specifications
Specifications
IgG2a, κ
G155-178
Alexa Fluor 647
Purified
Mouse
Mouse BALB/c IgG2a, κ
RUO
Primary
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Isotype Control
TNP-keyhole limpet hemocyanin
50μg
Cell Biology
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.