Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a Mouse anti-Mouse, Clone: C184, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221943250UG

Catalog No. NB004943


Add to cart

Description

Description

IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for .
Specifications

Specifications

IgG2a
C184
Unconjugated
Purified
IgG2a
250 ug
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Monoclonal
Mouse
Control
LYOPH
0.2 um filtered solution in PBS.
Mouse
Protein A or G purified
RUO
Isotype Control
Reconstitute with 1.0 ml PBS.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit