Learn More
IgG2a Mouse, BB700, Clone: G155-178, Isotype Control, BD Horizon™
Mouse Isotype Control
Manufacturer:  BD Biosciences 566419
Catalog No. BDB566419
Add to cart
Description
Description
IgG2a antibody specifically detects IgG2a in samples. It is validated for Flow Cytometry.
Specifications
Specifications
IgG2a
G155-178
BB700
Mouse
50 μg
Refrigerate (2 to 8°C)
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ