Learn More
IgG2a, κ, Mouse, PE, Clone: MOPC-173, Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Manufacturer:  BD Biosciences 565363
Catalog No. BDB565363
Add to cart
Description
Description
IgG2a, κ Monoclonal antibody specifically detects IgG2a, κ in samples. It is validated for Flow cytometry (Intracellular Staining)), Isotype control.
Specifications
Specifications
IgG2a, κ
MOPC-173
PE
Purified
None
50μg
Cell Biology
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
Mouse BALB/c IgG2a, κ
RUO
Primary
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.