Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgG2a Mouse, PE, Isotype Control, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Isotype Control

Manufacturer:  Invitrogen™ MG2A04

Catalog No. 50-112-1633


Add to cart

Specifications

Specifications

IgG2a
PE
Conjugated
IgG2a
ASR/IVD
Isotype Control
Flow Cytometry
PBS with 0.1% Sodium Azide
Mouse
0.5mL
Store at 2 - 8 °C.
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit