Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a Rabbit anti-Mouse, Biotin, Clone: RM219, Secondary Antibody, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP262008B
Description
IgG2a Monoclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
IgG2a | |
RM219 | |
Biotin | |
50% Glycerol/PBS with 1% BSA with 0.09% Sodium Azide | |
Rabbit | |
IgG | |
50 ug | |
Secondary | |
This antibody reacts to the Fc region of mouse IgG2a. No cross reactivity with mouse IgG1, IgG2b, IgG2c, IgG3, IgM, IgA, IgE, human IgG, or rat IgG. |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
Western Blot 0.5ug/mL - 2ug/mL, Flow Cytometry, ELISA 0.005ug/mL- 0.2ug/mL, Immunohistochemistry 0.5ug/mL - 2ug/mL, Immunoprecipitation, Immunocytochemistry 0.5ug/mL - 2ug/mL | |
Purified | |
Mouse IgG2a | |
Protein A purified | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title