Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a, κ Rat, Brilliant Ultraviolet 496, Clone: R35-95, Isotype Control, BD Horizon™

Rat Isotype Control

Manufacturer:  BD Biosciences 612951

Catalog No. BDB612951


Add to cart

Description

Description

IgG2a, κ Monoclonal isotype control is validated for Flow cytometry.
Specifications

Specifications

IgG2a, κ
R35-95
Brilliant Ultraviolet 496
Affinity Purified
Mouse Pooled Immunoglobulin
Affinity Chromatography
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50 μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit