Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a, κ Rat, Brilliant Ultraviolet 661, Clone: R35-95, Isotype Control, BD Horizon™
Rat Isotype Control
Supplier: BD Biosciences 612973
Description
Rat Isotype Control is validated for Flow Cytometry.Specifications
IgG2a, κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Pooled Immunoglobulin | |
50 μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
R35-95 | |
Brilliant Ultraviolet 661 | |
Rat | |
Affinity Purified | |
RUO | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction