Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a Rat, BUV563, Clone: R35-95, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 612922
Description
IgG2a Monoclonal is validated for Flow Cytometry, Control.Specifications
IgG2a | |
R35-95 | |
Brilliant Ultraviolet 563 | |
50 μg | |
Primary | |
IgG2a κ |
Flow Cytometry | |
0.2 mg/mL | |
Rat | |
RUO | |
Refrigerate (2 to 8°C) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction