Learn More
IgG2a κ Rat, RB780, Clone: R35-95, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568698
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.
Specifications
IgG2a κ | |
R35-95 | |
RB780 | |
Rat | |
50 μg | |
Isotype Control | |
IgG2a κ |
Flow Cytometry | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Mouse Pooled Immunoglobulin | |
RUO | |
Refrigerate (2°C to 8°C) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.