Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), PE, eBioscience™, Invitrogen™
Rat Isotype Control
$126.65 - $259.65
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9651
|
Invitrogen
12403181 |
50 μg |
Each
for $126.65
Save 9% Reg : |
|
|||||
50-112-9672
|
Invitrogen
12403182 |
100 μg |
Each for $189.00
|
|
|||||
50-186-38
|
Invitrogen
12403183 |
200 μg |
Each
for $259.65
Save 10% Reg : |
|
|||||
Description
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This monoclonal rat IgG2b kappa isotype control has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.Specifications
Rat IgG2b kappa | |
0.2 mg/mL | |
PE | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eB149/10H5 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.