Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b kappa, PE-Cyanine7, clone: eB149/10H5, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Rat Isotype Control

Manufacturer:  Invitrogen 25403181

 View more versions of this product

Catalog No. 50-112-9677


Add to cart

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: PE-Cy7 Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis.Applications Tested: Rat IgG2b K Isotype Control PE-Cyanine7 has been tested by flow cytometric analysis of mouse splenocytes. It should be used at the same concentration as the experimental antibody.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Rat IgG2b kappa
Control
0.2mg/mL
Liquid
IgG2, IgG2b, IgG2bk
IgG2b, kappa,kappa
Affinity chromatography
RUO
Not Applicable
Control,Flow Cytometry
eB149/10H5
PE-Cyanine7
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Rat
Monoclonal
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This Cyanine7 tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage,handling and experimental procedures.
Isotype Control
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.