Learn More
IgG2b kappa Mouse, Isotype Control, eFluor 660, eBioscience™ Invitrogen™
Mouse Isotype Control
Manufacturer:  LIFE TECHNOLOGIES 50473280
Encompass
Catalog No. 50-473-280
Add to cart
Description
Description
The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: Mouse IgG2b K Isotype Control eFluor 660 has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: Mouse IgG2b K Isotype Control eFluor 660 has been tested by flow cytometric analysis. This should be used at the same concentration as the experimental antibody. eFluor 660 is a replacement for Alexa Fluor 647. eFluor 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IgG2b kappa Mouse, Isotype Control, eFluor 660, eBioscience™ Invitrogen™ is validated for Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.
Specifications
Specifications
Mouse IgG2b kappa
Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Mouse
Affinity chromatography
RUO
Isotype Control
Liquid
eBMG2b
eFluor 660
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Not Applicable