Learn More
IgG2b Kpa ItCl Mouse, R718, Clone: 27-35, Isotype Control, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566952
Catalog No. BDB566952
Add to cart
Description
Description
This mouse IgG2b, κ isotype control is a monoclonal antibody, clone 27-35, that is specific for the dansyl (5-[dimethylamino] naphthalene-1-sulfonyl) hapten. The dansyl (DNS) hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing that demonstrated low background staining on a variety of mouse and human tissues.
Specifications
Specifications
IgG2b Kpa ItCl
27-35
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Isotype Control