Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b, κ Mouse, Brilliant Ultraviolet 737, Clone: 27-35, Isotype Control, BD Horizon™

Mouse Isotype Control

Manufacturer:  BD Biosciences 612768

Catalog No. BDB612768


Add to cart

Description

Description

IgG2b, κ Monoclonal isotype control is validated for Flow cytometry.
Specifications

Specifications

IgG2b, κ
27-35
Brilliant Ultraviolet 737
Affinity purified
IgG2b, κ
50 μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit