Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2b, Mouse, Clone: PC11, Isotype Control, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2219440.5MG
Description
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for .Specifications
IgG2b | |
PC11 | |
0.2 um filtered solution in PBS. | |
Protein A or G purified | |
Isotype Control | |
Mouse | |
Lyophilized |
Monoclonal | |
Unconjugated | |
Mouse | |
0.5 mg | |
Reconstitute with 1.0 mL PBS. | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title