Learn More
IgG2b Rat, BB700, Clone: R35-38, Isotype Control, BD Horizon™
Rat Isotype Control
Manufacturer:  BD Biosciences 566418
Catalog No. BDB566418
Add to cart
Description
Description
IgG2b antibody specifically detects IgG2b in samples. It is validated for Flow Cytometry.
Specifications
Specifications
IgG2b
R35-38
BB700
Rat
50 μg
Refrigerate (2 to 8°C)
Flow Cytometry
0.2 mg/mL
Purified
IgG2b, κ