Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG3, κ, Clone: J606, Isotype Control, BD

Manufacturer:  BD Biosciences 562482

Catalog No. BDB562482


Add to cart

Description

Description

Mouse IgG3 κ isotype control antibody is specific for the β-2 - 6-linked Fructosan. The transplantable plasmacytoma J606 was induced by intraperitoneal injection of mineral oils into BALB/c mice. This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20nm filter).

Host Species: Mouse
Clone: J606
Isotype: IgG3, κ

Flow Cytometry, Isotype Control

Specifications

Specifications

J606
PE-CF594
Mouse
100μg
0.2mg/ml
Affinity Purified
IgG3, kappa
4°C. Store in dark. Do not freeze.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit