Learn More
Athens Research And Technology IncSupplier Diversity Partner IGG3 HUMAN PLASMA 1MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 161609070731MG
Immunoglobulin G3, Human Normal Plasma: 1 MG; Liquid in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 170,000; EC= 1.36; Purity: > =95% by SDS-PAGE. Native; Prepared and purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-808