Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG3 Lambda Human, HRP, Isotype Control, Novus Biologicals™
Human Polyclonal Isotype Control
Supplier: Novus Biologicals NBP306871H
This item is not returnable.
View return policy
Description
IgG3 Lambda Polyclonal antibody specifically detects IgG3 Lambda in Human samples. It is validated for ELISA, ControlSpecifications
IgG3 Lambda | |
Polyclonal | |
PBS | |
Human myeloma serum | |
RUO | |
Store at 4°C in the dark. | |
IgG3 λ |
ELISA | |
HRP | |
Human | |
0.1 mL | |
Human | |
Isotype Controls |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction