Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG3 Rabbit anti-Human, Clone: RM119, Abnova™

Rabbit monoclonal antibody raised against native human IgG3.

Manufacturer:  Abnova Corporation MAB12820

Catalog No. 89-934-329


Add to cart

Description

Description

Specifications

Specifications

IgG3
RM119
Rabbit monoclonal antibody raised against native human IgG3.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
DKFZp686H11213/FLJ39988/FLJ40036/FLJ40253/FLJ40587/FLJ40789/FLJ40834/IgG3/MGC45809
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to the heavy chain of human IgG3. No cross reactivity with human IgG1, IgG2, IgG4, IgM, IgA, IgD, IgE, mouse IgG, rat IgG, or goat IgG.
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Unconjugated
ELISA (0.05-0.2 ug/mL for detection; 0.05-0.2 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IGHG3
Native purified human IgG3.
Protein A affinity purification
RUO
Primary
3502
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit