Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgGF(ab)2 Chicken, Isotype Control, Novus Biologicals™

Chicken Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1967712MG

Catalog No. NBP1967712


Add to cart

Description

Description

IgGF(ab)2 antibody specfically detects IgGF(ab)2 samples. It is validated for Western Blot, ELISA, SDS-Page.
Specifications

Specifications

IgGF(ab)2
2.0mg/mL
Western Blot, ELISA
Purified
IgG
2mg
Isotype Controls
Chicken
ELISA, Western Blot
Unconjugated
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Chicken
Ion exchange chromatography
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Chicken IgG, anti-Chicken IgG F(ab)2 and anti-Chicken Serum. No reaction was observed against anti-Chicken IgG F(c) or anti-Pepsin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit