Learn More
IgGF(ab)2 Hamster, Isotype Control, Novus Biologicals™
Hamster Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1967821MG
Catalog No. NBP1967821
Add to cart
Description
Description
IgGF(ab)2 antibody specfically detects IgGF(ab)2 samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgGF(ab)2
1.0mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
Purified
IgG
1mg
Isotype Controls
Golden Syrian Hamster
ELISA, Western Blot
Unconjugated
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Hamster
Ion exchange chromatography
Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Golden Syrian Hamster IgG, anti-Golden Syrian Hamster IgG F(ab)2 and anti-Golden Syrian Hamster Serum. No reaction was observed against anti-Golden Syrian Hamster IgG F(c) or anti-Pepsin.