Learn More
IgGFc Goat anti-Human, Agarose, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19726310MG
Catalog No. NBP19726310
Add to cart
Description
Description
IgGFc Polyclonal antibody specifically detects IgGFc in Human samples. It is validated for Immunoprecipitation.
Specifications
Specifications
IgGFc
Agarose
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Goat
IgG
10mg
Store at 4°C. Do not freeze.
Polyclonal
Immunoprecipitation
Immunoprecipitation
Purified
Human IgG Fc fragment
IgG purified
Immunology
Secondary
Human