Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHE, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548483

Catalog No. PA548483

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IGHE
Polyclonal
PBS with 0.09% sodium azide
P01854
Ig epsilon chain C region, IGHE
Rabbit
IgG
400μL
Store at 2° to 8°C for up to 6 months. For long term storage store at -20°C.
Primary
3497
Western Blot
Unconjugated
IGHE
Liquid
IGHE
KLH conjugated synthetic peptide between 246-275 amino acids from the Central region of human IGHE.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit