Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Heavy Chain Antibody (IG1707R), Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00 - $542.50
Specifications
Antigen | IGHG1 |
---|---|
Clone | IG1707R |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25314920
|
Novus Biologicals
NBP25314920UG |
20 μg |
Each for $301.00
|
|
|||||
NB253149100
|
Novus Biologicals
NBP253149100UG |
100 μg |
Each for $542.50
|
|
|||||
NB253149200
|
Novus Biologicals
NBP253149200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
IgG Heavy Chain Monoclonal specifically detects IgG Heavy Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGHG1 | |
0.2 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Immunology | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
3500 | |
Purified human Ig Gamma Chain | |
Primary | |
Store at 4C. |
IG1707R | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
immunoglobulin heavy constant gamma 1 (G1m marker) | |
IGHG1 | |
IgG κ | |
Protein A purified | |
75 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title