Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHG1 Rabbit anti-Human, PE/Cy5.5, Clone: IG1707R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254449PECY55

Catalog No. N254449PEC5


Add to cart

Description

Description

IGHG1 Monoclonal antibody specifically detects IGHG1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IGHG1
IG1707R
Flow Cytometry
Conjugated, Purified
IGHG1
Purified human Ig Gamma Chain
Protein A purified
Immunology
Primary
3500.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit