Learn More
Abnova™ IGHG4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003503T01
Catalog No. 89-948-509
Add to cart
Description
Description
Specifications
Specifications
293T
BC025985.1
pCMV-IGHG4 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
IGHG4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 52.14kDa
Transient overexpression cell lysate was tested with anti-IGHG4 antibody (Mfr. No. H00003503-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human