Sino Biological™ IGHG4 Recombinant Human Protein (108 Ser/Pro) Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human IgG4 Fc region (Glu 99- Lys 327, 108Ser/Pro)

Manufacturer: Sino Biological™ 13505HNAH25

 View more versions of this product

Catalog No. 13-505-HNAH25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 25.5kd
    Gene Symbol IGHG4
    Protein Tag Untagged
    Format Lyophilized
    Gene ID (Entrez) 3503
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Source Human
    Structural Form Truncated, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name IGHG4 Recombinant Human Protein (108 Ser/Pro)
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    Product Type IGHG4 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications

    Expressed from DNA sequence encoding human IgG4 Fc region (Glu 99- Lys 327, 108Ser/Pro)

    • Formulation: Lyopihilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit