Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgL Mouse anti-Human, PE/Atto594, Clone: LLC/1738, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254420PEATT594

Catalog No. N254420P594


Add to cart

Description

Description

IgL Monoclonal antibody specifically detects IgL in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IgL
LLC/1738
Flow Cytometry
Conjugated, Purified
IGL
Purified human full-length lambda light chain
Protein A or G purified
Adaptive Immunity, Immunology
Primary
3535.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
ig lambda-6 chain C region, IGL, IGLC6, immunoglobulin lambda gene cluster, immunoglobulin lambda locus, Lambda Light Chain, MGC88804
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit