Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGLL5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549022

Catalog No. PA549022


Only null left
Add to Cart

Description

Description

IGLL5 Polyclonal Antibody for Western Blot

This gene encodes one of the immunoglobulin lambda-like polypeptides. It is located within the immunoglobulin lambda locus but it does not require somatic rearrangement for expression. The first exon of this gene is unrelated to immunoglobulin variable genes; the second and third exons are the immunoglobulin lambda joining 1 and the immunoglobulin lambda constant 1 gene segments. Alternative splicing results in multiple transcript variants. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGLL5
Polyclonal
Unconjugated
IGLL5
anti HBs antibody light-chain Fab fragment; G lambda-1; Germline immunoglobulin lambda 1; IGL; IGLL5; IGLV; immunoglobin anti-granzymeB light chain variable region; immunoglobulin lambda like polypeptide 5; immunoglobulin lambda-3 surrogate light chain; immunoglobulin lambda-like polypeptide 5; omega light chain protein 14.1; VL-MAR
Rabbit
Antigen affinity chromatography, Protein A
RUO
100423062
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
B9A064
IGLL5
KLH conjugated synthetic peptide between 167-193 amino acids from the C-terminal region of human IGLL5.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.