Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM, BB515, Clone: G155-228, Isotype Control, BD
Supplier: BD Biosciences 564680
Description
Specifications
IgM | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
TNP-Keyhole Limpet Hemocyanin | |
50 μg | |
4°C, protect from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
G155-228 | |
Brilliant Blue 515 (BB515) | |
Mouse | |
Affinity Purified | |
Secondary | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction