Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ IgM (μ-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody with BSA and azide

IgM (μ-Heavy Chain) Ab-2, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Supplier:  Epredia™ RB1434A

 View more versions of this product

Catalog No. RB1434A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: IgM isolated from human serum of a patient with Waldenstrom's

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-2 recognizes the μ heavy chain of human immunoglobulins. Ab-2 does not cross-react with γ (IgG), α (IgA), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

IgM (µ-Heavy Chain) Ab-2
Polyclonal
Rabbit
1 mL
Immunoglobulins
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IgM isolated from human serum of a patient with Waldenstroms
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit