Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Mouse anti-Chicken, Turkey, APC, Clone: M-1, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 831011
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
IgM | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Flow Cytometry, ELISA | |
M-1 | |
APC | |
Mouse | |
RUO | |
Chicken, Turkey | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title