Learn More
IgM Mouse anti-Human, DyLight 350, Clone: MA2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107092UV
Catalog No. NB1107092UV
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgM Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
IgM
MA2
Western Blot, ELISA
Purified
IGHM
IgM from human serum.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Mu-chain of human IgM, Fab - region localized epitope.
Western Blot, ELISA
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Mouse
IgG2b
0.2 ml
B Cell Development and Differentiation Markers, Immunology
Primary
3507
Human