Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Rabbit anti-Mouse, Clone: RM109, Abnova™
Rabbit monoclonal antibody raised against native mouse IgM.
Supplier: Abnova Corporation MAB12772
Specifications
IgM | |
Monoclonal | |
Unconjugated | |
ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user. | |
AI326478/IGHM/Igh-M/Igh6/Igm/MGC107036/MGC107507/muH | |
Rabbit | |
Protein A affinity purification | |
RUO | |
16019 | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
RM109 | |
Rabbit monoclonal antibody raised against native mouse IgM. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Igh-6 | |
Native purified IgM from mouse hybridoma. | |
100 μg | |
Primary | |
This antibody reacts to mouse IgM. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, human IgG, rat IgG, or goat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction