Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF21 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549304

Catalog No. PA549304

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein which has two immunoglobulin (Ig) domains and is a member of the immunoglobulin superfamily. Proteins in this superfamily are usually found on or in cell membranes and act as receptors in immune response pathways. [provided by RefSeq, Sep 2011] Orthologs mouse all
Specifications

Specifications

IGSF21
Polyclonal
PBS with 0.09% sodium azide
Q96ID5
Immunoglobulin superfamily member 21, IgSF21, IGSF21
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84966
Western Blot
Unconjugated
IGSF21
Liquid
IGSF21
KLH conjugated synthetic peptide between 87-115 amino acids from the N-terminal region of human IGSF21.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit