Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4A/SynCAM1/CADM1 Rat anti-Human, Alexa Fluor™ 350, Clone: 41N3D3, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP214841AF350

Catalog No. NP214841A30


Add to cart

Description

Description

IGSF4A/SynCAM1/CADM1 Monoclonal antibody specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IGSF4A/SynCAM1/CADM1
41N3D3
Western Blot
Conjugated, Purified
CADM1
A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody.
Protein G purified
Growth and Development, Neuronal Cell Markers, Neuroscience
Primary
23705.0
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
Rat
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit