Learn More
BioVision IncSupplier Diversity Partner Ready-to-use IHC kit (Biotin free), One-Step HRP Polymer anti-Mouse, Rat and Rabbit IgG (H+L) with DAB: Highly sensitive, biotin/avidin free IHC/ICC kit to stains membrane, cytoplasmic and nuclear antigens. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K40550
Immunohistochemistry (IHC)/Immunocytochemistry (ICC) is the localization of antigens in tissue sections/cells by the use of labeled antibodies as specific reagents through antigen-antibody interactions that are visualized by a marker such as fluorescent dye, enzyme, radioactive element or colloidal gold. Several IHC techniques are commonly used such as labeled biotin secondary antibody streptavidin-peroxidase, HRP anti-HRP, ABC catalyzed signal amplification, polymer system (one or two steps) etc. to detect antigens in tissues and cells. BioVision’s Ready-to-use IHC kit employs polymer technology that provides increased sensitivity and detection. IBSC One-Step anti-mouse, Rat and Rabbit IgG (H+L) is a biotin/avidin free system that stains membrane, cytoplasmic and nuclear antigens. It provides the user with a rapid and easy to use IHC detection system. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9739