Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IHR-Cy3, >95%, Tocris Bioscience™

Click to view available options
Quantity:
1mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent fluorescent Smo antagonist

Potent fluorescent Smo antagonist (IC50 = 100 nM).

Non-fluorescent analog also available.

Quantity 1mg
Linear Formula C58H63Cl3N6O10S2
Formula Weight 1174.64
Grade >95%
Chemical Name or Material (2-[3-[1-(6-[[7-[[4-Chloro-3-[[4-(2,5-dichlorobenzamido)phenyl]carbamoyl]phenyl]amino]-7-oxoheptylam

Recommended Storage : Store at −20°C