Learn More
IKA™ RV05 Basic Series Rotary Evaporators
Manufacturer:  IKA™ 8017900
Catalog No. 14-259-276
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
17.7 in.
900 mm
12 kg
220/240 or 100/120 V
520 mm
46 to 260 rpm
Digital
220 to 240 / 100 to 120 / 100 to 115V, 1133W, 50/60Hz
Asynchronous
1133 w Input x 1000 w Output
IP 20
DIN EN 60529
50/60 Hz
450 mm
35.4 in.
26.4 lb.
20.4 in.
1000 mL/hr.
Ambient to 225°C
1200 cm2
Set of glassware RV 06.2 with vertical condenser (space-saving), drive RV 05 basic, telescopic stand RV 05.3 and boss head clamp R 271.
41°F to 104°F
Diagonal
Without Controller
Rotary Evaporator
RV 05 basic 1-B