Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IkB-alpha Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793835PLATES

 View more versions of this product

Catalog No. NB009806


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IkB-alpha.
Specifications

Specifications

12.5 pg/mL
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL mouse anti-IkB-alpha monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-IkB-alpha monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 22 ng of recombinant IkB-alpha.
  • RUO
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IkB-alpha.
    IkB-alpha
    Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
    Human
    NFKBIA
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.8 ng/mL is recommended.
    HRP
    4792
    Antibody Pairs
    12.5-800 pg/mL
    Storage is content dependent.
    Mouse/Rabbit
    5 Plates
    Sandwich ELISA
    alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit