Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$401.35 - $550.85

Specifications

Antigen IkB-beta (p Ser23)
Host Species Rabbit
Isotype IgG
Gene Alias beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Target Species Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP254762 View Documents Novus Biologicals
NBP254762
100 uL Each for $401.35
Add to cart
 
NBP254761 View Documents Novus Biologicals
NBP254761
0.1 mg Each for $550.85
Add to cart
 
Description

Description

IkB-beta p Ser23 Polyclonal antibody specifically detects IkB-beta p Ser23 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IkB-beta (p Ser23)
IgG
Human, Mouse, Rat
RUO
Affinity Purified
Primary
Rabbit IgG in phosphate buffered saline pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Rabbit
beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Store at -20 degreesC.
Polyclonal

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit