Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKB α Rabbit anti-Human,Mouse, Unconjugated, Clone: 6O8, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen IKB α
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB114425MI
View Documents
MilliporeSigma
ZRB114425UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11444XMI
View Documents
MilliporeSigma
ZRB11444X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

IKB α Monoclonal antibody specifically detects IKB α in Human,Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

IKB α
Unconjugated
Rabbit
NF-kappa-B inhibitor alpha;I-kappa-B-alpha;IkB-alpha;IkappaBalpha;Major histocompatibility complex enhancer-binding protein MAD3
His-tagged recombinant fragment corresponding to 143 amino acids from the N-terminal half of human I-kappa-B-alpha.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
NFKBIA, IKBA, MAD3, NFKBI
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit