Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKBKAP Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $436.00
Specifications
Antigen | IKBKAP |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 -1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
IKBKAP Polyclonal specifically detects IKBKAP in Human samples. It is validated for Western Blot.Specifications
IKBKAP | |
Western Blot 1:500 -1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
8518 | |
Amino acid residues 148-161 IHQDDFGESKFITV of human IKAP. Cross reactivity: sequence 92% identical to rat and 85% identical to mouse. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
158 kDa |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
ELP1 | |
IgG | |
Affinity Purified | |
Reacts with the N terminus of IKAP. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title