Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKB (IKK Beta) Recombinant Human Protein

Encompass

IKBKB (IKK Beta) Recombinant Human Protein

$491.00

Specifications

Accession Number NP_001547
Expression System Baculovirus
Gene Alias IKBKB (IKK Beta)
KinaseFamily IKK Kinase Family
Molecular Weight 114 kDa
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3836
Encompass
View Documents Promotion Details
LIFE TECHNOLOGIES
PV3836
10μg
Each for $491.00
Add to cart
 
Description

Description

Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex.
Specifications

Specifications

NP_001547
IKBKB (IKK Beta)
114 kDa
GST Tag
Baculovirus
IKK Kinase Family
Kinase Assay
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit