Learn More
IKBKE Mouse anti-Human, Clone: OTI3F10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807633
Catalog No. 50-166-8382
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IKBKE is a noncanonical I-kappa-B kinase (IKK) that is essential for regulating antiviral signaling pathways. IKBKE has also been identified as a breast cancer oncogene and is amplified and overexpressed in over 30% of breast carcinomas and breast cancer cell lines.
Specifications
Specifications
IKBKE
Monoclonal
Unconjugated
IKBKE
Lyophilized
IKBKE
Human recombit protein fragment corresponding to amino acids 474-716 of human IKBKE produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI3F10
PBS with 8% trehalose and no preservative; pH 7.3
Q14164
IKK-E, IKK-i, IKKE, IKKI
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9641